Learn More
Tenak™ Comfort Rack for Chest Freezers, 75 mm Box Height, with Boxes
Comfort Rack suitable for use with all chest freezer models.
Brand: Tenak™ TE24544B
Additional Details : Weight : 2.50000kg
Description
- Racks with spring locks mounded on each shelf to secure the cryoboxes
- Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
- Measure the inside height of your freezer when choosing the right rack
- Constructed of stainless steel
- Includes standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Spring Locks | |
Stainless Steel | |
140 x 140 x 707 mm |
Boxes (9) | |
Chest Frseezers | |
75 mm (Box) |