Learn More
Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height, with Boxes
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand:  Tenak™ TE24302B
Additional Details : Weight : 3.27000kg
 View more versions of this product
Product Code. 15210784
  • 2625.00 SEK / Each
Estimated Shipment: 24-07-2024
to see stock.
Add to basket
Description
Description
  • In the comfort rack, the cryoboxes are placed in drawers
  • Increases the flexibility and reduces the operating weight compared to the rack types
  • Measure the inside height and depth of you freezer when choosing the right rack
  • Racks can be used with all freezer models
  • Constructed of stainless steel
  • Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications
Specifications
100 mm (Box)
Upright Freezers
2 x 4
562 x 140 x 214 mm
Boxes (8)