Learn More
Tenak™ Comfort Rack for Upright Freezers, 100 mm Box Height, with Boxes
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24296B
Additional Details : Weight : 3.40000kg
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
- Include standard white cardboard boxes with 9 x 9 cell dividers
Specifications
100 mm (Box) | |
Upright Freezers | |
3 x 3 |
424 x 140 x 315 mm | |
Boxes (9) | |
Stainless Steel |