Learn More
Tenak™ Comfort Rack for Upright Freezers, 50 mm Box Height
Comfort Rack designed to ensure a more ergonomical operating process when working with samples.
Brand: Tenak™ TE24226
Additional Details : Weight : 3.60000kg
Description
- In the comfort rack, the cryoboxes are placed in drawers
- Increases the flexibility and reduces the operating weight compared to the rack types
- Measure the inside height and depth of you freezer when choosing the right rack
- Racks can be used with all freezer models
- Constructed of stainless steel
Specifications
50 mm (Box) | |
Boxes (18) | |
Stainless Steel |
Upright Freezers | |
6 x 3 |