Läs mer
Invitrogen™ COMP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA595547
Beskrivning
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human HepG2 whole cell, human Hacat whole cell, human CACO-2 whole cell, human 293T whole cell, rat cartilage tissue, mouse cartilage tissue. IHC: mouse knee-joint tissue, rat knee-joint tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
COMP is a noncollagenous extracellular matrix protein. It consists of five identical glycoprotein subunits, each with EGF like and calcium binding (thrombospondin like) domains. Oligomerization results from formation of a five stranded coiled coil and disulfides. Binding to other ECM proteins such as collagen appears to depend on divalent cations. Mutations can cause the osteochondrodysplasias pseudochondroplasia (PSACH) and multiple epiphyseal dysplasia (MED).
Specifikationer
| COMP | |
| Polyclonal | |
| Unconjugated | |
| COMP | |
| Cartilage oligomeric matrix protein; cartilage oligomeric matrix protein (pseudoachondroplasia, epiphyseal dysplasia 1, multiple); cartilage oligomeric matrix protein(pseudoachondroplasia, epiphyseal dysplasia 1, multiple); Comp; EDM1; EPD1; MED; MGC131819; MGC149768; PSACH; pseudoachondroplasia (epiphyseal dysplasia 1, multiple); putative cartilage oligomeric matrix protein; THBS5; Thrombospondin5; Thrombospondin-5; TSP 5; TSP5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12845, 1311, 25304 | |
| -20°C | |
| Lyophilized |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin), Western Blot, Immunohistochemistry (Paraffin) | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P35444, P49747, Q9R0G6 | |
| COMP | |
| E.coli-derived human COMP recombinant protein (Position: Q505-A757). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.