Läs mer
Buerkle™ Compact Lever Pump
Handla allt Bürkle Produkter
Material:
Polypropylene
Beskrivning
- Polypropylene, seals made of NBR
- Pump capacity up to about 500 ml per stroke
- Discharge tube can be rotated 360°
- With integrated barrel screw connection for G2 in. thread
Specifikationer
Specifikationer
| Flödeshastighet | 500 mL/stroke |
| Pumpslangens längd | 100 cm |
| Motståndsegenskaper | Alkalis, Detergent, Weak Acid |
| Typ | Hand Pump |
| Material | Polypropylene |
| Anslutningstyp | G 2 in. |