Läs mer
Complement C3c Antibody (4), Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-49651
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Complement C3c Monoclonal specifically detects Complement C3c in Human samples. It is validated for Immunoprecipitation.
Specifikationer
| Complement C3c | |
| Monoclonal | |
| 0.1 mg/mL | |
| Immunoprecipitation 1:10-1:500 | |
| C3 | |
| The monoclonal antibody 4 recognizes a conformational epitope in C3c, C3b and iC3b. | |
| 0.025 mg | |
| Immunology, Innate Immunity, Neuronal Cell Markers, Neuroscience | |
| 718 | |
| Store at 4°C. |
| Immunoprecipitation | |
| 4 | |
| Unconjugated | |
| AHUS5, ARMD9, ASP, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 1, complement C3, complement component 3, complement component C3, CPAMD1acylation-stimulating protein cleavage product | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Korrigering av produktinnehåll
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering