missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Complement Component C3aR Antibody (620613) [DyLight 755], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB10417Z
4480.00 SEK valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Complement Component C3aR Monoclonal antibody specifically detects Complement Component C3aR in Mouse samples. It is validated for Flow Cytometry, CyTOF
Specifications
Complement Component C3aR | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
620613 | |
Flow Cytometry, CyTOF-ready | |
AZ3B, AZ3BC3a anaphylatoxin chemotactic receptor, C3a-R, C3AR, C3AR1, C3ARcomplement component 3 receptor 1, C3R1, complement component 3a receptor 1, Complement Component C3a R, Complement Component C3aR, HNFAG09 | |
Chinese hamster ovary cell line CHO-derived transfected with mouse C3aR, Accession # O09047 | |
0.1 mL | |
GPCR | |
719 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction