missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ CompletePAK Sterile Kit with 5mL Serum Vials, 20mm Stopper, and Flip Seal

Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application

14337.00 SEK - 16044.00 SEK

Specifications

Closure Size 20mm
Sterility Sterile
Material Glass
No. per Case 200
Product Line Wheaton CompletePAK
View More Specs

Products 6
Product Code Brand Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
Product Code Brand Closure Type Cap Style Volume (Metric) Price Quantity & Availability  
16277300 View Documents DWK Life Sciences
W990006-CP
Ultra Pure Serum Stopper, Blue Flip Seal 5 mL 14337.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
16287300 View Documents DWK Life Sciences
W990007-CP
Omni Flex 3G Straight Plug Stopper, Blue Flip Seal 5 mL 15634.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
16297300 View Documents DWK Life Sciences
W990008-CP
Igloo Lyo Stopper, Blue Flip Seal 5 mL 16044.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
16207310 View Documents DWK Life Sciences
W990009-CP
Ultra Pure Serum Stopper, Red Flip Seal 5 mL 14337.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
16217310 View Documents DWK Life Sciences
W990010-CP
Omni Flex 3G Straight Plug Stopper, Red Flip Seal 5 mL 15634.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
16227310 View Documents DWK Life Sciences
W990011-CP
Igloo Lyo Stopper, Red Flip Seal 5 mL 16044.00 SEK
Pack of 200 View alternative products
Save Up to 
»
Estimated Shipment
01-10-2019
Add to basket
 
Description

Description

  • Type I glass serum vials
  • Sterile 5mL vials
  • Range of sterile stoppers and seals
  • Each component is particulate cleaned, dehydrogenated, and sterilized certified to USP <788>, <85> & <71>
  • All components are Container Closure Integrity (CCI) certified
  • Each kit has 220 stoppers and seals

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications

Specifications

20mm
Glass
Wheaton CompletePAK
Sterile
200
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified