Läs mer
Concanamycin A, Tocris Bioscience™
Beskrivning
Concanamycin A is a specific inhibitor of V-type (vacuolar) H+-ATPase that displays > 2000-fold selectivity over other H+-ATPases (IC50 values are 9.2, > 20000, > 20000 and > 20000 nM for yeast V-type, F-type, P-type H+-ATPases and porcine P-type Na+,K+-ATPase respectively). Blocks cell surface expression of virus envelope glycoproteins without affecting synthesis and exhibits cytotoxicity in several cell lines.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Concanamycin A |
| Typ | Concanamycin A |
| CAS | 80890-47-7 |
| Kvantitet | 10 μg |
| Molekylformel | C46H75NO14 |
| MDL-nummer | MFCD00210037 |
| Synonym | (3Z,5E,7R,8R,9S,10S,11R,13E,15E,17S,18R)-18-[(1S,2R,3S)-3-[(2R,4R,5S,6R)-4-[[4-O-(Aminocarbonyl)-2,6-dideoxy-β-D-arabino-hexopyranosyl]oxy]tetrahydro-2-hydroxy-5-methyl-6-(1E)-1-propenyl-2H-pyran-2-yl]-2-hydroxy-1-methylbutyl]-9-ethyl-8, |
| InChI-nyckel | DJZCTUVALDDONK-HQMSUKCRSA-N |
| LEDER | CC[C@H]1[C@@H](O)[C@H](C)C\C(C)=C\C=C\[C@H](OC)[C@H](OC(=O)\C(OC)=C\C(\C)=C\[C@@H](C)[C@H]1O)[C@@H](C)[C@@H](O)[C@H](C)[C@@]1(O)C[C@@H](O[C@H]2C[C@@H](O)[C@H](OC(N)=O)[C@@H](C)O2)[C@H](C)[C@H](O1)\C=C\C |
| IUPAC-namn | (2R,3S,4R,6R)-6-{[(2R,4R,5S,6R)-2-[(2S,3R,4S)-4-[(2R,3S,4E,6E,9R,10S,11S,12R,13R,14E,16Z)-11-ethyl-10,12-dihydroxy-3,17-dimethoxy-7,9,13,15-tetramethyl-18-oxo-1-oxacyclooctadeca-4,6,14,16-tetraen-2-yl]-3-hydroxypentan-2-yl]-2-hydroxy-5-methyl-6-[(1E)-prop-1-en-1-yl]oxan-4-yl]oxy}-4-hydroxy-2-methyloxan-3-yl carbamate |
| Visa mer |