Learn More
Lenz Laborglasintrumente Condensers with Receiver Acc. To Twisselmann
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 05420106
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10070251
  • 5861.80 SEK / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket
Description
Description
  • Condensers with receiver
  • Acc. to Twisselmann
  • Made of DURAN™ tubing
Specifications
Specifications
100 to 250 mL
Borosilicate Glass 3.3
NS 45/40