missing translation for 'onlineSavingsMsg'
Learn More

Conductivity solution 100,000μS/cm, Fisher chemical™

Catalog Number p-7083755
Click to view available options
Quantity:
500 mL
1000 mL
Packaging:
Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Packaging Bottle
Quantity 500 mL
Type Conductivity Solution
Conductivity 100,000μS/cm at 25°C
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'