Learn More
Lenz Laborglasintrumente Conical Joint Clips Type Keck™
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
356.16 SEK - 615.86 SEK
Product Code | Brand | For Use With (Equipment) | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Color | Price | Quantity & Availability | ||||
10050271
|
Lenz Laborglasintrumente
01103745 |
For ground joint NS 45/40 | Brown |
615.86 SEK
Pack of 10 |
Estimated Shipment: 23-05-2024
Log in to see stock. |
||||
10569941
|
Lenz Laborglasintrumente
01103712 |
For ground joint NS 12/21 | Violet |
356.16 SEK
Pack of 10 |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
Description
- For conical ground joints
- Material: POM, outstanding chemical resistance
- Operating temperature up to approx. 150 °C
Specifications
POM |