Läs mer
Conivaptan Hydrochloride, TRC
Produktkod. 30012165
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012165 10 mg
30011453 25 mg
30005162 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012165 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC38470010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 168626-94-6
Molekylformel C32 H26 N4 O2 . Cl H
Molekylvikt (g/mol) 535.04
Synonym N-[4-[(4,5-Dihydro-2-methylimidazo[4,5-d][1]benzazepin-6(1H)-yl)carbonyl]phenyl]-[1,1'-biphenyl]-2-carboxamide Hydrochloride (1:1), [1,1'-Biphenyl]-2-carboxamide, N-[4-[(4,5-dihydro-2-methylimidazo[4,5-d][1]benzazepin-6(1H)-yl)carbonyl]phenyl]-, monohydrochloride (9CI), Conivaptan hydrochloride, Vaprisol, YM 087, conivaptan monohydrochloride
IUPAC-namn N-[4-(2-methyl-4,5-dihydro-1H-imidazo[4,5-d][1]benzazepine-6-carbonyl)phenyl]-2-phenylbenzamide;hydrochloride
LEDER Cl.Cc1nc2CCN(C(=O)c3ccc(NC(=O)c4ccccc4c5ccccc5)cc3)c6ccccc6c2[nH]1
Specifikationer
Kemiskt namn eller material Conivaptan Hydrochloride
Kvantitet 10 mg
InChI formel InChI=1S/C32H26N4O2.ClH/c1-21-33-28-19-20-36(29-14-8-7-13-27(29)30(28)34-21)32(38)23-15-17-24(18-16-23)35-31(37)26-12-6-5-11-25(26)22-9-3-2-4-10-22;/h2-18H,19-20H2,1H3,(H,33,34)(H,35,37);1H
Formel vikt 534.1823