Läs mer
Conjugated Linoleic Acid, 90%(Mixture of Isomers), TRC
Produktkod. 30367393
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30367393 500 mg
30358826 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30367393 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC685000500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 22880-03-1
Molekylformel C18H32O2
Molekylvikt (g/mol) 280.4
Synonym Conjugated Linoleic Acid, 90% ( Isomers)
IUPAC-namn (10E,12E)-octadeca-10,12-dienoic acid
LEDER CCCCC\C=C\C=C\CCCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material Conjugated Linoleic Acid (Mixture of Isomers)
Kvantitet 500 mg
InChI formel InChI=1S/C18H32O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h6-9H,2-5,10-17H2,1H3,(H,19,20)/b7-6+,9-8+
Formel vikt 280.2402