Learn More
Lenz Laborglasintrumente™ Connecting Spring for Ground Joints
For alu rings with hooks to connect conical ground joints safely
Brand:  Lenz Laborglasintrumente™ 01102840
Additional Details : Weight : 0.00100kg
Product Code. 10510132
  • 360.40 SEK / Pack of 10
Estimated Shipment: 30-04-2024
to see stock.
Add to basket
Description
Description
  • Stainless Steel
  • Two rings
Specifications
Specifications
10
Two rings
40 mm length
Stainless Steel
Alu rings to connect conical ground joints