Learn More
WTW™ Connection Cable for Depth Probes
For use with WTW™ MPP IDS depth probes with waterproof plug head.
Brand:  WTW™ 903851
Additional Details : Weight : 0.30000kg
 View more versions of this product
Product Code. 15230396
  • 1514.72 SEK / Each
Estimated Shipment: 11-07-2024
to see stock.
Add to basket
Description
Description
  • Also useful for plug head sensors
  • Available in a range of lengths
  • Supplied individually
Specifications
Specifications
Connection Cable
3 m