missing translation for 'onlineSavingsMsg'
Learn More

Connexin 43 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen Connexin 43
Concentration 0.33 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16825305
View Documents Promotion Details
Proteintech
26980-1-AP-20UL
20 μL
1656.73 SEK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16815305
View Documents Promotion Details
Proteintech
26980-1-AP-150UL
150 μL
4721.67 SEK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 17-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene is a member of the connexin gene family. The encoded protein is a component of gap junctions, which are composed of arrays of intercellular channels that provide a route for the diffusion of low molecular weight materials from cell to cell. The encoded protein is the major protein of gap junctions in the heart that are thought to have a crucial role in the synchronized contraction of the heart and in embryonic development. A related intronless pseudogene has been mapped to chromosome 5. Mutations in this gene have been associated with oculodentodigital dysplasia and heart malformations.
Specifications

Specifications

Connexin 43
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse
P17302, P23242
14609, 2697
Connexin 43 Fusion Protein Ag25533
Primary
-20°C
GJA1
0.33 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Connexin 43, CX43, DFNB38, Gap junction alpha 1 protein, GJA1, GJAL, ODDD
Gja1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications