missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beskrivning
Connexin 47 Monoclonal Antibody for Western Blot
This gene encodes a member of the gap junction protein family. The gap junction proteins are membrane-spanning proteins that assemble to form gap junction channels that facilitate the transfer of ions and small molecules between cells. According to sequence similarities at the nucleotide and amino acid levels, the gap junction proteins are divided into two categories, alpha and beta. Mutations in this gene cause X-linked Charcot-Marie-Tooth disease, an inherited peripheral neuropathy. Alternatively spliced transcript variants encoding the same protein have been found for this gene.
Specifikationer
Specifikationer
| Antigen | Connexin 47 |
| Användningsområden | Western Blot |
| Klassificering | Monoclonal |
| Klona | 391CT6.4.3 |
| Koncentration | 2 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 0.09% sodium azide; pH 7.4 |
| Gen | GJC2 |
| Genaccessionsnr. | Q5T442 |
| Gene Alias | B230382L12Rik; connexin 47 type A; connexin 47 type C; connexin 47 type D; connexin-46.6; Connexin-47; CX46.6; Cx47; CXG2; gap junction alpha-12 protein; Gap junction gamma-2 protein; gap junction membrane channel protein alpha 12; gap junction protein gamma 2; gap junction protein, alpha 12; gap junction protein, chi 2; gap junction protein, gamma 2; gap junction protein, gamma 2, 47kDa; GJA12; Gjc2; HLD2; LMPH1C; PMLDAR; SPG44 |
| Visa mer |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?
