Learn More
Contec™ PROSAT™ PS-911 Presaturated Wipes
PROSAT™ wipes are an effective, efficient and convenient method of cleaning process tools and other equipment. PROSAT wipes are also ideal for wiping articles prior to pass through.
Brand: Contec™ PS-911
Additional Details : CAS Number : 67-63-0 Weight : 16.00000kg Transport : UN number : 3175 Chem class : 4.1 Pack group : II
Description
- Presaturated with IPA and DI water blends in flat, resealable pouches individually lot-numbered for traceability
- These specially designed pouches preserve cleanliness and solvent saturation levels
- Solvent blend meets SEMI specifications for metals and contaminants
- ISO Class 5 compatible
- 30 wipes/pouch, 50 pouches/case