Learn More
Contec™ PROSAT™ Presaturated Nonwoven Wipes
Use these wipes to clean laminar flow workbenches, cleanroom surface and equipment. Contec™ PROSAT™ Presaturated Nonwoven Wipes feature task-specific saturation level of the solvent or disinfectant required to do the job.
Brand:  Contec™ PS-911
Additional Details : CAS Number : 64-17-5 Weight : 16.00000kg Transport : UN number : 3175 Chem class : 1 Pack group : II
 View more versions of this product
Product Code. 15476474
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +46 31 352 32 00 for assistance
View alternative products »
Description
Description
  • Wipe substrate designed for the application
  • Compatible with ISO Class 5 (FED-STD-209E Class 100/M3.5) environments
  • Ideal for wiping articles prior to pass through
  • Made of meltblown polypropylene or cellulose/polyester
  • Effective and convenient for wipe-downs of process tools and other equipment
  • Choose spunlaced 100% polyester wipers for improved cleanliness, meltblown polypropylene wipes for improved durability, or economical hydroentangled polyester/cellulose wipers
  • Solvent blend meets SEMI specifications for metals and contaminants
Specifications
Specifications
ISO Class 5
Process tools and other equipment
70% IPA/ 30% DI Water
23 cm
28 cm
PROSAT™ Presaturated Wipes
11 x 9 in. (28 x 23 cm)
30 wipes/bag
Yes
Ideal para la limpieza de rutina de instrumentos y equipos