Learn More
Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads
Product Code. 18886610 Shop All Contec Products
Change view
Click to view available options
Size:
Small
Medium
Large
For Use With (Equipment):
12 in. Medium Frame
14 in. Large Frame
7.5 in. Regular Frame
Unit Size:
Each
Pack of 32
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size For Use With (Equipment) Unit Size
18886610 Small 7.5 in. Regular Frame Each
15964970 Medium 12 in. Medium Frame Each
16615524 Large 14 in. Large Frame Pack of 32
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18886610 Supplier Contec™ Supplier No. VKMOPIR
  • ONLINE EXCLUSIVE PRICE
    11395.00 SEK
    9083.75 SEK / Paket med 1
    Save 2311.25 SEK
    20% Av
 Restricted Item
Beräknad leverans: 15-02-2027
Use with VertiKlean™ Wall Washing System which is designed for cleaning vertical surfaces. VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
  • Lightweight, polyester, disposable
  • Ideal for applying disinfectants and wet cleaning vertical surfaces, ceilings, and walls in controlled environments; reaching awkward corners and ceilings with a complete range of motion
  • Absorbent foam carries solvents and disinfectants to the surface; polyester fabric offers excellent abrasion resistance and helps distribute the liquid uniformly on the surface being cleaned
  • Heads are laser cut to seal edges, further limiting potential contamination in the mopping process
  • Textured surface option is ideal for applications requiring additional scrubbing performance
  • Contain no cellulose; compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
Order Info
Aluminum handle with mop frame available separately (Cat. No. 19-038-555).
TRUSTED_SUSTAINABILITY
Specifications
Product Type Mop Head
Sterility Sterile
Autoclavable Autoclavable
Size Small
Disposable Reusable
For Use With (Equipment) 7.5 in. Regular Frame
For Use With (Application) For applying disinfectants and wet cleaning ceilings and walls in controlled environments
Material Knitted Polyester, Polyester Foam