Learn More
SymLine Continuous Level Control
Product Code. 15988462
Change view
Click to view available options
Product Type:
Level Control
Unit Size:
Each
Product Code. Product Type Unit Size
15988462 Level Control Each
1 options
Product Code. 15988462 Supplier SymLine Supplier No. 502042
  • ONLINE EXCLUSIVE PRICE
    21645.00 SEK
    17274.21 SEK / Paket med 1
    Save 4370.79 SEK
    20% Av
Beräknad leverans: 03-12-2026
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!
  • Automatic warning for 5 different filling levels freely adjustable - accuracy of up to ± 0.5 mm
  • The control box provides 5 freely configurable outlets to control external devices
  • The compact design and independent power supply allow simple handling and installation, also outside switch cabinets
  • Disc sensor for level control from non-conductive material - easy to install and adjustable in height
  • Rod Sensor and Canister with sleeve - Removable sleeve for easy maintenance and cleaning with canister made from electrostatic conductive PE-HD material
  • Specifications
    Product Type Level Control