Läs mer
Convallatoxin, TRC
Produktkod. 30134342
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134342 5 mg
18032150 1 mg
18062650 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134342 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC6850755MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 508-75-8
Molekylformel C29 H42 O10
Molekylvikt (g/mol) 550.64
Synonym Card-20(22)-enolide, 3-[(6-deoxy-α-L-mannopyranosyl)oxy]-5,14-dihydroxy-19-oxo-, (3β,5β)-, Convallatoxin (6CI,7CI,8CI), (3β,5β)-3-[(6-Deoxy-α-L-mannopyranosyl)oxy]-5,14-dihydroxy-19-oxocard-20(22)-enolide, Convallaotoxin, Convallaton, Convallatoxine, Convallatoxoside, Convallotoxin, Corglycon, Corglycone, Corglykon, Korglykon, NSC 407808, Strophanthidin α-L-rhamnoside
IUPAC-namn (3S,5S,8R,9S,10S,13R,14S,17R)-5,14-dihydroxy-13-methyl-17-(5-oxo-2H-furan-3-yl)-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthrene-10-carbaldehyde
LEDER C[C@@H]1O[C@@H](O[C@H]2CC[C@]3(C=O)[C@H]4CC[C@]5(C)[C@H](CC[C@]5(O)[C@@H]4CC[C@]3(O)C2)C6=CC(=O)OC6)[C@H](O)[C@H](O)[C@H]1O
Specifikationer
Kemiskt namn eller material Convallatoxin
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C29H42O10/c1-15-22(32)23(33)24(34)25(38-15)39-17-3-8-27(14-30)19-4-7-26(2)18(16-11-21(31)37-13-16)6-10-29(26,36)20(19)5-9-28(27,35)12-17/h11,14-15,17-20,22-25,32-36H,3-10,12-13H2,1-2H3/t15-,17-,18+,19-,20+,22-,23+,24+,25-,26+,27-,28-,29-/m0/s1
Formel vikt 550.2778
Procent renhet >95%