Learn More
3M™ Cool White Protective Coverall 4532+
Catalog Number p-7160333
1365.00 SEK valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
3M™ Protective Coverall 4532+ has a unique specialist material treatment, which offers enhanced liquid hold out with added oil and alcohol repellency.
  • Unique liquid repellency with oil and alcohol resistance material treatment
  • Soft and strong multi-layered 52 gsm SSMMS for strong performance
  • Made from super breathable SSMMS non-woven for effective release of body heat and moisture and reduced heat stress
  • 3 panel hood, reinforced triangular gusset panel, 2-way zip and storm flap with sealable tape, elasticated back and ankles to better freedom of movement
  • Anti-static coating EN 1149-5 reduces static discharge and the risk of sparks
  • EN 1073-2 nuclear particulate hold out
  • Knitted cuffs and elastic waist and ankles for a better fit and freedom of movement
Product Type Protective Coveralls
Size Small
Material SSMMS
Color White
Waist Style Elastic
Cuff Style Knit
Closure Type 2-way Zipper
Certifications/Compliance CE category III , EN 1073-2 radioactive particles , EN 1149-5 - electrostatic properties , EN 13034: 2005 + A1: 2009 Type 6 liquid splash , EN ISO 13982-1: 2004 + A1: 2010 Type 5 harmful particles