Läs mer
Copper Acetoarsenite (Technical Grade), TRC
Produktkod. 30353951
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30353951 1 g
1 options
Produktkod. 30353951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC6852551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Copper Acetoarsenite
CAS 12002-03-8
Molekylformel 2 C2 H3 O2 . 6 As O2 . 4 Cu
Kvantitet 1 g
Synonym Paris green, copper acetoarsenite
InChI formel InChI=1S/2C2H4O2.6AsHO2.4Cu/c2*1-2(3)4;6*2-1-3;;;;/h2*1H3,(H,3,4);6*(H,2,3);;;;/q;;;;;;;;4*+2/p-8
LEDER [Cu+2].[Cu+2].[Cu+2].[Cu+2].CC(=O)[O-].CC(=O)[O-].[O-][As]=O.[O-][As]=O.[O-][As]=O.[O-][As]=O.[O-][As]=O.[O-][As]=O
IUPAC-namn tetracopper;oxoarsinite;diacetate
Molekylvikt (g/mol) 1013.79
Formel vikt 1011.2136