missing translation for 'onlineSavingsMsg'
Learn More

Copper, Granular, 20 to 30 Mesh, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog Number p-5391691
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

High quality chemicals for laboratory and specialized industrial use

Quantity 500 g
Formula Weight 63.55
Grade BAKER ANALYZED™ Reagent
Mesh Size 20 to 30 Mesh
Packaging Poly Bottle
Form Granular
Chemical Name or Material Copper
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'