missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) cyanide, 99%, extra pure, Thermo Scientific Chemicals

Catalog Number 10454985
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
This item is not returnable. View return policy
544-92-3
CCuN
89.56
MFCD00010975
DOBRDRYODQBAMW-UHFFFAOYSA-N
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
11009
copper(1+);cyanide
[C-]#N.[Cu+]
This item is not returnable. View return policy
544-92-3
CCuN
89.56
MFCD00010975
DOBRDRYODQBAMW-UHFFFAOYSA-N
copper i cyanide, cupricin, copper +1 cyanide, copper cyanide cu cn, cyanocopper, rcra waste number p029, cucn, rcra waste no. p029, cyano copper
11009
copper(1+);cyanide
[C-]#N.[Cu+]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY
Melting Point 474.0°C
Color Beige-White to Green
Physical Form Powder
Quantity 5 kg
Assay Percent Range 98.5% min. (ex Cu)
Linear Formula CuCN
Fieser 05,166; 06,146; 09,124; 16,97
Merck Index 15, 2651
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in acids and alkalies,practically insoluble in alcohol
Formula Weight 89.56
Percent Purity 99%
Grade Extra Pure
Packaging Plastic Drum
Specific Gravity 2.92
Density 2.9200g/mL
Chemical Name or Material Copper(I) cyanide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(I) cyanide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H310-Fatal in contact with skin.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
  • EUH032-Contact with acids liberates very toxic gas.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'