Learn More
Copper(I) oxide, 99% (metals basis), Thermo Scientific Chemicals
Catalog Number p-4407431
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
1317-39-1
Cu2O
MFCD00010974
1317-39-1
Cu2O
MFCD00010974
Used in antifouling, marine paint, brazing paste, antifungal treatment for plant seeds. Used as a hydrogen scavenger in copper brass and bronze and high grade lubricants. Finds uses in glass and ceramic pigments, ferrite magnets, catalyst in organic synthesis, dyes and dye intermediates and as catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Boiling Point 1800°C (decomposition)
Melting Point 1235°C
Color Purple-Red
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (Metal basis)
Solubility Information It is insoluble in water and organic solvents.
Formula Weight 143.08
Odor Odorless
Refractive Index 2.705
Sensitivity Air and moisture sensitive
Appearance Purple-red powder
Density 6
Chemical Name or Material Copper(I) oxide