missing translation for 'onlineSavingsMsg'
Learn More

Copper(Ii) Acetate Monohydrate, ACS Reagent, 98.0 to 102.0%, Honeywell Fluka™

Catalog Number 15682050
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O
This item is not returnable. View return policy
6046-93-1
C4H8CuO5
199.65
MFCD00149570
NWFNSTOSIVLCJA-UHFFFAOYSA-L
copper ii acetate monohydrate, cupric acetate monohydrate, copper diacetate monohydrate, copper acetate monohydrate, copper 2+ acetate, monohydrate, acetic acid, copper 2+ salt, monohydrate, verdigris, copper acetate hydrate, diacetoxycopper hydrate, copper acetate, hydrate
165397
copper;diacetate;hydrate
O.[Cu++].CC([O-])=O.CC([O-])=O

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Melting Point 115°C
Color Blue to Green
Quantity 2.5 kg
Linear Formula Cu(CO2CH3)2 · H2O
UN Number UN1759
Beilstein 3730548
Formula Weight 199.65g/mol
Percent Purity ≥98%
Grade ACS Reagent
Packaging Poly Bottle
Appearance Powder/Crystals
Density 1.88g/cu.cm; Vapor Density: 6.8 (vs air)
Chemical Name or Material Copper(Ii) Acetate Monohydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'