missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Nitrate Hemi(Pentahydrate), Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 6 x 1 kg |
Linear Formula | Cu(NO3)2 · 2.5H2O |
UN Number | 3085 |
Formula Weight | 232.59g/mol |
Percent Purity | ≥98% |
Chemical Name or Material | Copper(Ii) Nitrate Hemi(Pentahydrate) |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'