missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) picolinate, 98%, Thermo Scientific™

Catalog Number p-7077660
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
14050-01-2
C12H8CuN2O4
307.75
GNWVUSJJFONIKF-UHFFFAOYSA-L
copper ii picolinate, unii-96e596pypp, copper picolinate, copper, bis picolinato, copper picolinate anhydrous, copper, bis 2-pyridinecarboxylato-kappan1,kappao2, copper 2+ ion dipicolinate, copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2
14963587
copper;pyridine-2-carboxylate
C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]
This item is not returnable. View return policy
14050-01-2
C12H8CuN2O4
307.75
GNWVUSJJFONIKF-UHFFFAOYSA-L
copper ii picolinate, unii-96e596pypp, copper picolinate, copper, bis picolinato, copper picolinate anhydrous, copper, bis 2-pyridinecarboxylato-kappan1,kappao2, copper 2+ ion dipicolinate, copper,bis 2-pyridinecarboxylato-kn1,ko2-, hydrate 1:2
14963587
copper;pyridine-2-carboxylate
C1=CC=NC(=C1)C(=O)[O-].C1=CC=NC(=C1)C(=O)[O-].[Cu+2]
Quantity 1 g
Assay Percent Range 97.5% min. (Iodometry)
Formula Weight 307.75
Percent Purity 98%
Infrared Spectrum Authentic
Loss on Drying 0.02
Packaging Glass Bottle
Chemical Name or Material Copper(II) picolinate
compliance-icons compliance-icons
Product Identifier
  • Copper(II) picolinate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'