missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate, anhydrous, 98%, Thermo Scientific Chemicals

Catalog Number p-7021587
Click to view available options
Quantity:
250 g
1000 g
5000 g
7758-98-7
CuO4S
159.602
MFCD00010981
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
24462
CHEBI:23414
copper;sulfate
[O-]S(=O)(=O)[O-].[Cu+2]
7758-98-7
CuO4S
159.602
MFCD00010981
ARUVKPQLZAKDPS-UHFFFAOYSA-L
copper sulfate, copper ii sulfate, cupric sulfate anhydrous, copper sulphate, copper 2+ sulfate, blue stone, copper monosulfate, copper ii sulfate, anhydrous, hylinec, trinagle
24462
CHEBI:23414
copper;sulfate
[O-]S(=O)(=O)[O-].[Cu+2]

Copper(II) sulfate, 98%, CuO4S, CAS Number-7758-98-7, copper 2+ sulfate, copper ii sulfate, blue stone, copper monosulfate, copper sulphate, trinagle, copper sulfate, hylinec, cupric sulfate anhydrous, copper ii sulfate, anhydrous, 1000g, CHEBI:23414, 3.6, Hazard Class: 9; Packaging Group: III

Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling's solution and Benedict's solution to test for reducing sugars. Also, used to test protein in Biuret test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Copper(II)sulfate is used in electroplating and in mining industries. It is used as a dehydrating agent for forming and manipulating acetal groups. In analytical chemistry, it is used in Fehling′s solution and Benedict′s solution to test for reducing sugars. Also, used to test protein in Biuret test.

Solubility
Soluble in water and methanol. Insoluble in ethanol.

Notes
Incompatible with strong bases, metals, alkali metals and powdered metals.
TRUSTED_SUSTAINABILITY
Melting Point >560°C
Quantity 250 g
Linear Formula CuSO4
UN Number UN3288
Merck Index 14,2653
Solubility Information Soluble in water and methanol. Insoluble in ethanol.
Formula Weight 159.6
Percent Purity 98%
Odor Odorless
Sensitivity Hygroscopic
Density 3.6
Chemical Name or Material Copper(II) sulfate, anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'