missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(Ii) Sulfate Pentahydrate, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
500 g
2.5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | CuSO4 · 5H2O |
UN Number | 3077 |
IUPAC Name | copper(2+) pentahydrate sulfate |
Formula Weight | 249.69g/mol |
Percent Purity | ≥98% |
Grade | ACS Reagent |
Vapor Pressure | 7.3mmHg (25°C) |
Chemical Name or Material | Copper(Ii) Sulfate Pentahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'