missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Copper(II) sulfate pentahydrate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., 99-102%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
500 g
1 kg
50 kg
6 x 1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Quantity | 100 g |
Linear Formula | CuSO4 · 5H2O |
UN Number | 3077 |
Formula Weight | 249.69g/mol |
Percent Purity | 99 to 102% |
Grade | ACS/puriss. p.a. |
Vapor Pressure | 7.3mmHg (25°C) |
Chemical Name or Material | Copper(Ii) Sulfate Pentahydrate, Reag. Ph. Eur. reag. ISO |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'