Läs mer
Copper(II) Sulfate, TRC
Produktkod. 30134727
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134727 5 g
30136363 10 g
30004318 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134727 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC7250405G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7758-98-7
Molekylformel Cu . O4 S
Molekylvikt (g/mol) 159.61
Synonym Cupric sulfate, Copper(2+) sulfate (1:1) (ACI), Baijunling, Black blue stone, Blue Copper, Blue stone, Blue vitriol, Bluestone, Copper monosulfate, Copper sulfate, Copper sulfate (1:1), Copper sulfate (CuSO4), Copper(2+) sulfate, Copper(II) sulfate, Cuivrol, Cupric sulfate anhydrous, Copper (II) sulphate anhydrous, Cupric sulphate, CuPRiX20, Delcup, EarthTec, Hylinec, Incracide 10A, Incracide E 51, MAC 570, Mastercop, Monocopper sulfate, Reducer Cu, Roman vitriol, Sulfuric acid, copper(2+) salt (1:1), Sulfuric acid, monocopper(1+) salt, Upinorg, XWHGS 005
IUPAC-namn copper;sulfate
LEDER [Cu+2].[O-]S(=O)(=O)[O-]
Specifikationer
Kemiskt namn eller material Copper sulfate
Kvantitet 5 g
InChI formel InChI=1S/Cu.H2O4S/c;1-5(2,3)4/h;(H2,1,2,3,4)/q+2;/p-2
Formel vikt 158.8813