Learn More
Copper(II) trifluoromethanesulfonate, 99%, Thermo Scientific Chemicals
Description
Copper(II) trifluoromethanesulfonate is a mild lewis acid. It is used as catalyst which promotes dehydration of alcohols and diols to alkenes at ambient temperatures. It is widely used to generate carbenoid species from ?-diazo esters and ketones, via in situ reduction to the Cu(I) species. It is also promotes the reaction between diazo esters and imines to give aziridines. It catalyzes syn-selective aldol condensation of (Z)-silyl enol ethers with aldehydes, Friedel-Crafts alkylation, acylation reactions of aromatics and addition of trimethylsilyl cyanide to carbonyl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | (decomposition) |
Color | Blue to White |
Physical Form | Powder |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 4028198 |
Solubility Information | Soluble in water. |
Formula Weight | 361.68 |
Percent Purity | ≤99% |
Sensitivity | Hygroscopic |
Show More |
RUO – Research Use Only