Läs mer
Copper Oxalate Hemihydrate, TRC
Produktkod. 30359214
Change view
Kvantitet:
25 g
Produktkod. Kvantitet
30359214 25 g
1 options
Produktkod. 30359214 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC68536025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Copper Oxalate Hemihydrate
CAS 55671-32-4
Molekylformel C5 H12 N4
Kvantitet 25 g
Synonym Copper(II) Oxalate, Cupric Oxalate, [Ethanedioato(2-)-O,O'] Copper Hemihydrate, [Ethanedioato(2-)-kO1,kO2] Copper Hemihydrate
InChI formel InChI=1S/C5H12N4/c1-9(2)5(8)3-4(6)7/h8H,3H2,1-2H3,(H3,6,7)
LEDER CN(C)C(=N)CC(=N)N
IUPAC-namn 3-N,3-N-dimethylpropanediimidamide
Molekylvikt (g/mol) 128.176
Formel vikt 128.106