Läs mer
Copper Pyrithione, TRC
Produktkod. 30138504
Change view
Kvantitet:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138504 100 mg
30135736 500 mg
30005426 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC685425100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 154592-20-8
Molekylformel 2C5 H4 N O S . Cu
Molekylvikt (g/mol) 315.86
IUPAC-namn copper;1-oxidopyridine-2-thione
LEDER [Cu+2].[O-]N1C=CC=CC1=S.[O-]N2C=CC=CC2=S
Specifikationer
Kemiskt namn eller material Copper Pyrithione
Kvantitet 100 mg
InChI formel InChI=1S/2C5H4NOS.Cu/c2*7-6-4-2-1-3-5(6)8;/h2*1-4H;/q2*-1;+2
Formel vikt 314.9323