Läs mer
Coprostan-3-ol, TRC
Produktkod. 30009640
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009640 100 mg
30005166 10 mg
30003824 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009640 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC685450100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 360-68-9
Molekylformel C27 H48 O
Molekylvikt (g/mol) 388.67
Synonym Cholestan-3-ol, (3β,5β)-, 5β-Cholestan-3β-ol (8CI), (3β,5β)-Cholestan-3-ol, 3β-Hydroxy-5β-cholestanol, 5β-Coprostanol, Coprostan-3-ol, Coprostan-3β-ol, Coprostanol, Coprosterol, Koprosterin, Koprosterol, NSC 18175, NSC 5060, Stercorin
IUPAC-namn (3S,5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-3-ol
LEDER CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material Coprostan-3-ol
Kvantitet 100 mg
InChI formel InChI=1S/C27H48O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h18-25,28H,6-17H2,1-5H3/t19-,20-,21+,22+,23-,24+,25+,26+,27-/m1/s1
Formel vikt 388.3705