missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COPS2/TRIP15 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | COPS2/TRIP15 |
---|---|
Concentration | 0.49 mg/mL |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887703
|
Proteintech
10969-2-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877703
|
Proteintech
10969-2-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 25-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Product No. 10969-2-AP is a rabbit polyclonal antibody raised against the full-length of human COPS2. The MW of this protein is 52 kDa, and this antibody specifically recognizes the 52 kDa protein.
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predomitly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of the tricarboxylic acid cycle. Each isozyme is a heterotetramer that is composed of two alpha subunits, one beta subunit, and one gamma subunit. The protein encoded by this gene is the beta subunit of one isozyme of NAD(+)-dependent isocitrate dehydrogenase. Three alternatively spliced transcript variants encoding different isoforms have been described for this gene.Specifications
COPS2/TRIP15 | |
Immunofluorescence, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
P61201, P61202 | |
12848, 9318 | |
COPS2/TRIP15 Fusion Protein Ag1415 | |
Primary | |
-20°C | |
COPS2 |
0.49 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ALIEN, Alien homolog, COPS 2, COPS2, CSN2, SGN2, Signalosome subunit 2, TR interacting protein 15, TRIP 15, TRIP15 | |
Cops2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |