missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COQ6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12481-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
COQ6 (Coenzyme Q10 monooxygenase 6) is also named as CGI-10 and belongs to the ubiH/COQ6 family. It is an evolutionarily conserved flavin-dependent monooxygenase required for biosynthesis of coenzyme Q10, or ubiquinone. It has 3 isoforms with the molecular mass of 51 kDa, 43 kDa and 49 kDa. In inner ear, Coq6 is expressed in spiral ganglion, stria vascularis cells, and spiral ligament. Defects in COQ6 are the cause of coenzyme Q10 deficiency, primary, type 6 (COQ10D6).Specifications
COQ6 | |
Polyclonal | |
Unconjugated | |
COQ6 | |
CGI 10, COQ6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
217707, 299195, 51004 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q68FU7, Q8R1S0, Q9Y2Z9 | |
COQ6 | |
COQ6 Fusion Protein Ag3164 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |