Läs mer
Corning™ BioCoat™ Analyssystem
Beskrivning
Innehåller
24-Multiwell™ Insert plate, specialized media to perform 24 assays, feeder tray, and lid.
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
Specifikationer
Specifikationer
| Färg | Clear |
| Kulturområde | 0.3 cm2 |
| Sterilitet | Non-sterile |
| Ytbehandling | Collagen |
| Typ | Cellkulturinsatssystem |
| Räkna väl | 24 |
| För användning med (applikation) | Rapid differentiation of Caco 2 cells in vitro for robotic screening of prospective pharmaceuticals for oral bioavailability and absorption |
| Inkluderar | 24 multiwell insert plate, specialized media to perform 24 assays, feeder tray and lid |
| Porstorlek | 1 μm |
| Kvantitet | 1/Cs. |