Learn More
Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water
High quality water for use in cell culture applications. Corning™ Cell Culture Grade Water Tested to USP/EP Sterile Water are used in preparation of purification and chromatography buffers.
12545.00 SEK - 18515.00 SEK
Specifications
For Use With (Application) USP/EP certified for injection purposes
Product Type Cell Culture Grade Water
Quantity 1 x 100L
View More Specs
Alert:
Product should not be used as an injectable or in the preparation of injectables.
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15343661 View Documents Corning™
25-065-LG
1 x 100L 12545.00 SEK
100 L View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
15353661 View Documents Corning™
25-065-LH
1 x 200L 18515.00 SEK
200 L View alternative products
Save Up to 
»
Estimated Shipment
06-05-2020
Add to basket
Description
Description
  • Tested to USP monograph for sterile water for injection (WFI)
  • Ideal for the preparation of reagents, rinsing glassware and plasticware, and other molecular biology applications where RNase, DNase, and Protease-free water is requiredFree of any added components
  • No toxic agents, including DEPC, are used, eliminating possible interferences with enzymatic reactions
  • Endotoxin levels ≤0.025EU/mL
  • Available in product sizes ranging from 20L to 200L
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
Specifications
Specifications
USP/EP certified for injection purposes
Cell Culture Grade Water
1 x 100L