Learn More
Corning™ Support Rod
For Use With: Models PC-400, PC-410, PC-420, PC-400D, PC-410d, PC-420D, PC-600D, PC-610d, PC-620D
Brand:  Corning™ 440129
Additional Details : Weight : 0.50000kg
Product Code. 10727338
  • 348.98 SEK / Each
Estimated Shipment: 25-10-2024
to see stock.
Add to basket
Description
Description
Corning offers a stainless steel, two piece, 18 (0.8cm) diameter that consists of two 9 rods that screw together. It screws into the base of Corning hot plate and stirrer models PC-400/410/420, PC-400D/410D/420D and PC-600D/610D/620D.
Specifications
Specifications
0.313 in.
18 in.
Rod
Models PC-400, PC-410, PC-420, PC-400D, PC-410d, PC-420D, PC-600D, PC-610d, PC-620D
45.7 cm