Läs mer
Corticosterone, Lipomed
Beskrivning
Corticosterone
Specifikationer
Specifikationer
| Kemiskt namn eller material | Corticosterone |
| CAS | 50-22-6 |
| Analyt- eller komponentnamn | Corticosterone |
| Frakt skick | Room Temperature |
| Molekylformel | C21 H30 O4 |
| Synonym | Pregn-4-ene-3,20-dione, 11,21-dihydroxy-, (11β)-, Corticosterone (8 CI), 11,21-Dihydroxyprogesterone, 11-Hydroxycorticoaldosterone, 11β,21-Dihydroxypregn-4-ene-3,20-dione, Reichstein's substance H, Reichstein's substance H, Reichstein's substance H, Reichstein's substance H, Reichstein's substance H, Reichstein's substance H, Reichstein's substance H |
| InChI formel | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| LEDER | C[C@]12 C[C@H](O)[C@H]3 [C@@H](CCC4=CC(=O)CC[C@]34 C)[C@@H]1 CC[C@@H]2 C(=O)CO |
| IUPAC-namn | (8 S,9 S,10 R,11 S,13 S,14 S,17 S)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-3-one |
| Industry Type | Pharmaceutical |
| Visa mer |