Läs mer
Corticosterone, MedChemExpress
Produktkod. 16474617
Encompass_Preferred
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
16474617 50 mg
1 options
Produktkod. 16474617 Leverantör Medchem Express Leverantörsnummer HYB161850mg
MedChemExpress Corticosterone (17-Deoxycortisol) is an orally active and adrenal cortex-produced glucocorticoid, which plays an important role in regulating neuronal functions of the limbic system (including hippocampus, prefrontal cortex, and amygdala).
Corticosterone increases the Rab-mediated AMPAR membrane traffic via SGK-induced phosphorylation of GDI. Corticosterone also interferes with the maturation of dendritic cells and shows a good immunosuppressive effect.
Specifikationer
Kemiskt namn eller material Corticosterone
CAS 50-22-6
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C21H30O4
Kvantitet 50 mg
Synonym 17-Deoxycortisol 11β,21-Dihydroxyprogesterone Kendall's compound B
Löslighetsinformation DMSO : 100 mg/mL (288.63 mM; Need ultrasonic) ∣Ethanol : 14.29 mg/mL (41.25 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (1.93 mM; Need ultrasonic)
LEDER C[C@@]12[C@@H](C(CO)=O)CC[C@@]1([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@@]3([H])[C@@H](O)C2)=O
Molekylvikt (g/mol) 346.46
Formel vikt 346.46
Procent renhet 95.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation