Läs mer
Cortienic Acid, TRC
Produktkod. 30351153
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30351153 100 mg
1 options
Produktkod. 30351153 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC695800100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Cortienic acid
CAS 3597-45-3
Molekylformel C20 H28 O5
Kvantitet 100 mg
Synonym Androst-4-ene-17-carboxylic acid, 11,17-dihydroxy-3-oxo-, (11β,17α)-, Androst-4-ene-17β-carboxylic acid, 11β,17-dihydroxy-3-oxo- (6CI,7CI,8CI), (11β,17α)-11,17-Dihydroxy-3-oxoandrost-4-ene-17-carboxylic acid, 11β,17-Dihydroxy-3-oxo-androst-4-ene-17β-carboxylic acid, 11β,17α-Dihydroxy-17β-carboxyandrost-4-en-3-one, 11β,17α-Dihydroxyandrost-4-en-3-one-17β-carboxylic acid, 17β-Carboxy-11β,17α-dihydroxy-4-androsten-3-one, Cortienic acid, 11β,17α-Dihydroxy-3-oxo-4-androsten-17β-carboxylic Acid
InChI formel InChI=1S/C20H28O5/c1-18-7-5-12(21)9-11(18)3-4-13-14-6-8-20(25,17(23)24)19(14,2)10-15(22)16(13)18/h9,13-16,22,25H,3-8,10H2,1-2H3,(H,23,24)/t13-,14-,15-,16+,18-,19-,20-/m0/s1
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)O)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC-namn (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-17-carboxylic acid
Molekylvikt (g/mol) 348.43
Formel vikt 348.1937