missing translation for 'onlineSavingsMsg'
Läs mer

Cortisol 21-Benzoate, TRC

Produktkod. 30361485
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30361485 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30361485 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC69631025MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material Cortisol 21-Benzoate
CAS 74669-84-4
Molekylformel C28H34O6
Kvantitet 25 mg
Synonym (11beta)-21-(Benzoyloxy)-11,17-dihydroxy-pregn-4-ene-3,20-dione, 21-Benzoyl Cortisol
InChI formel InChI=1S/C28H34O6/c1-26-12-10-19(29)14-18(26)8-9-20-21-11-13-28(33,27(21,2)15-22(30)24(20)26)23(31)16-34-25(32)17-6-4-3-5-7-17/h3-7,14,20-22,24,30,33H,8-13,15-16H2,1-2H3/t20-,21-,22-,24+,26-,27-,28-/m0/s1
LEDER C[C@]12CCC(=O)C=C1CC[C@H]3[C@@H]4CC[C@](O)(C(=O)COC(=O)c5ccccc5)[C@@]4(C)C[C@H](O)[C@H]23
IUPAC-namn [2-[(8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl]-2-oxoethyl] benzoate
Molekylvikt (g/mol) 466.57
Formel vikt 466.2355
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.