Läs mer
Cortisone 17-Valerate (>90%), TRC
Produktkod. 30138390
Change view
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138390 500 mg
30137313 250 mg
30007172 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138390 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC696550500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 136370-31-5
Molekylformel C26H36O6
Molekylvikt (g/mol) 444.56
Synonym 21-Hydroxy-17-[(1-oxopentyl)oxy]pregn-4-ene-3,11,20-trione
IUPAC-namn [(8S,9S,10R,13S,14S,17S)-17-(2-hydroxyacetyl)-10,13-dimethyl-3,11-dioxo-1,2,6,7,8,9,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
LEDER O=C1C[C@@]2(C)[C@](CC[C@@]2(OC(CCCC)=O)C(CO)=O)([H])[C@]3([H])CCC4=CC(CC[C@]4(C)[C@]31[H])=O
Specifikationer
Kemiskt namn eller material Cortisone 17-Valerate
Kvantitet 500 mg
InChI formel InChI=1S/C26H36O6/c1-4-5-6-22(31)32-26(21(30)15-27)12-10-19-18-8-7-16-13-17(28)9-11-24(16,2)23(18)20(29)14-25(19,26)3/h13,18-19,23,27H,4-12,14-15H2,1-3H3/t18-,19-,23+,24-,25-,26+/m0/s1